Drukuj
Spec-Tec™ Stretch M104 Sterile Glove Liners
Dostawca: WELLS LAMONT
M104EU-S M104EU-M M104EU-L M104EU-XL M104EU-XXL
112-6616EA 2120 PLN
112-6616 112-6615 112-6614 112-6617 112-6618
Spec-Tec™ Stretch M104 Sterile Glove Liners
Rękawice
Sterile and reusable cut-resistant glove liners.
  • For use in cleanroom environments
  • Cut-resistant
The M104 sterile cut-resistant stretch glove is constructed using latex-free HPPE fibers. An EN 388 rating of 4241 provides level 2 cut resistance. Each pair is individually wrapped around tissue paper in a secure sterile package. The M104 is an ideal glove in industries such as pharmaceutical, medical, and organ tissue processing.
Certyfikaty: EN 388 4241.
Sposób pakowania: Gloves are packaged in a box of 10 pairs. Each pair is sealed in its own sterile pouch.