Drukuj
Bizflame™ 88/12 FR89 Flame Resistant Shirts
Dostawca: Portwest
FR89NAR3XL FR89NAR4XL FR89NAR5XL FR89NAR6XL FR89NARL FR89NARM FR89NARS FR89NARXL FR89NARXXL
PRTWFR89NAR3XLEA 304 PLN
PRTWFR89NAR3XL PRTWFR89NAR4XL PRTWFR89NAR5XL PRTWFR89NAR6XL PRTWFR89NARL PRTWFR89NARM PRTWFR89NARS PRTWFR89NARXL PRTWFR89NARXXL
Bizflame™ 88/12 FR89 Flame Resistant Shirts
Odzież robocza
This stylish shirt is lightweight and comfortable. Maximum protection is guaranteed with permanent flame resistance and ARC2 protection against electric arc.
  • Dual hazard protection
  • Guaranteed flame resistant for life of garment
  • 2 front pockets, pen division on left pocket
  • Button cuffs for a secure and comfortable fit
  • Shirt tail hem stays tucked in
  • 40+ UPF rated fabric to block 98% of UV rays
Made from Bizflame 88/12 fabric: 88% Cotton, 12% Nylon.
Certyfikaty: ASTM F1959/F1959M-12, ATPV 8,2 cal./cm² (HAF 69,1%), ASTM F1506-10A, NFPA 70E, NFPA 2112.