N,O-Bis(trimethylsilyl)trifluoroacetamide with 1% trimethylchlorosilane
Dostawca: Thermo Fisher Scientific
|
Niebezpieczeństwo
|
Synonim:
BSTFA+TMCS (99:1)
Packaged under argon in resealable ChemSeal™ bottles.
packaged under Argon in resealable ChemSeal™ bottles
CAS No.: 25561-30-2
Molecular Formula: C8H18F3NOSi2
Formula Weight: 257.40
Storage Temperature: 2°C to 8°C
Flash Point: 23°(73°F)
Physical Form: Liquid
Density: 0.97
Refractive Index: 1.3840
Sensitivity: Moisture Sensitive
Appearance: Colourless to pale yellow
Melting Point: -10°
Boiling Point: 145-147°
Beilstein Reference: 3607620
UN No.: 2924
MDL No.: MFCD00008269
Formuła:
C₈H₁₈F₃NOSi₂ Temp. Wrzenia: 145…147 °C Temp. topnienia: –10 °C Gęstość: 0,97 Temp. Zapłonu: 23 °C (73 °F) Temperatura przechowywania: Lodówka |
Numer MDL:
MFCD00008269 Numer CAS: 25561-30-2 Numer WE (dawny EINECS): 247-103-9 Jednostka: 2924 ADR: 3,III |