Portelite™ Rapid Fluorimetric Endotoxin Detection Kits
Dostawca: AAT BIOQUEST
60008
60009
AATB60008EA
2050
PLN
AATB60008
AATB60009
Portelite™ Rapid Fluorimetric Endotoxin Detection Kits
Zestawy do oznaczeń life science
Testy komórkowe
Lipopolysaccharide (LPS), also known as endotoxin, is the major component of the outer membranes of Gram-negative bacteria.
- Sensitive detection
- Compatible with multiple sample types
- Strong and stable fluorescent signal
Uwaga: For research use only.