Human Recombinant Protein (from Wheat Germ), C9orf72
Dostawca: Abnova
H00203228-P01
H00203228-P012
ABNOH00203228-P01EA
1890
PLN
ABNOH00203228-P01
ABNOH00203228-P012
Human Recombinant Protein (from Wheat Germ), C9orf72
Białka
Human C9orf72 full-length ORF (NP_060795.1, 1 a.a. - 481 a.a.) recombinant protein with GST tag at N-terminal.
Storage Buffer: 50 mM Tris-HCI, 10 mM reduced; Glutathione, pH = 8,0 in the elution buffer.