Drukuj
Koncentratory odśrodkowe, Vivacon® 500, Sartorius
Dostawca: Sartorius
VN01H21 VN01H02 VN01H22 VN01H31 VN01H32 VN01H41 VN01H42 VN01H91 VN01H01 VN01H92 VN01H82
518-0002EA 735 PLN
518-0002 518-0001 518-0003 518-0004 518-0005 518-0006 518-0007 518-0008 518-0000 518-0009 518-0056
Koncentratory odśrodkowe, Vivacon® 500, Sartorius
Filtry Filtry i koncentratory wirówkowe
For samples up to 500 µl; membrane: Regenerated cellulose Hydrosart®. Vivacon® 500 centrifugal concentrators have been designed for DNA and protein concentration and rebuffering. They feature a horizontal membrane design that is preferred by some users for reasons of consistency with previously used procedures and reproducibility.
  • Optimal performance with very dilute samples, example forensic specimens
  • Low binding materials; high recoveries even from diluted samples
  • Re-spin possibility; complete and highly reproducible sample recovery
  • Volume graduation and the molecular weight cut-off printed on individual devices
* Starter pack.