Drukuj
EDGE® 48-160 Protective Leather Gloves
Dostawca: Ansell
48-160/10 48-160/9
ANSE48-160/10EA 207 PLN
ANSE48-160/10 ANSE48-160/9
EDGE® 48-160 Protective Leather Gloves
Rękawice
Protective leather gloves, granting superb mechanical performance, comfort and grip.
  • Palm, full forefinger and fingertips in split leather for added safety
  • Reinforced back with elastic wristband makes it a comfortable protective gloves
  • Good flexibility and abrasion resistance; durable and dexterous gloves
  • Heightened protection
  • Elevated comfort
  • Increased dexterity and durability
  • Cut and sewn construction
Featuring palm, full forefinger and fingertips in split leather, EDGE® 48-160 industrial gloves are solid and robust, making them ideal for mediumweight applications.
These hand protection gloves possess a reinforced back with an elasticated band at the wrist, which enhances user comfort.
EDGE® 48-160 high-dexterity leather gloves also deliver good flexibility and are suitable for a diverse range of tasks in the automotive, landscaping, warehousing and construction industries.
Recommended for assembling metal parts and components, handling of metal sheets, assembly and inspection of components, general maintenance and finishing works.
Primary industries: Automotive, landscaping, warehousing and construction.
Certyfikaty: Category II, EN 388, ISO 21420:2020, 2122A and REACH compliant.
Sposób pakowania: Six pairs in a bag; 12 bags in a carton.