Drukuj
Over-Trousers, 3000
Dostawca: Portwest
FB31NARXXL FB31NARL FB31NARXL FB31NARS FB31NARM
PRTWFB31NARXXLEA 2910 PLN
PRTWFB31NARXXL PRTWFB31NARL PRTWFB31NARXL PRTWFB31NARS PRTWFB31NARM
Over-Trousers, 3000
Odzież robocza
Designed to protect the lower body, Solar FB31 over-trousers are constructed using a Nomex®/Lenzing® FR viscose fabric. The design offers high freedom of movement. Features include a low-cut crotch seam, non detachable shoulder braces, side elastic and bellow front fly.
  • Protection against radiant, convective and contact heat
  • Waterproof and breathable with taped seams to prevent water penetration
  • Concealed hook and loop fastening for easy closure
  • Adjustable shoulder strap for added comfort
  • Ergonomic quick release buckle
  • Anti-wicking strip at hem
  • Sew on flame resistant industrial wash tape
  • Half elasticated waist for a secure and comfortable fit
  • CE-CAT III
Colour: Navy
Weather conditions: Dry, indoor workwear
Certyfikaty: EN 469 Level 2.