Drukuj
Whizard® Silver Talon Cut-Resistant Gloves
Dostawca: WELLS LAMONT
134527 134528 134529 134525 134526 135747
112-6368EA 512 PLN
112-6368 112-6369 112-6370 112-6366 112-6367 112-6371
Whizard® Silver Talon Cut-Resistant Gloves
Rękawice
Offers a very high level of cut resistance in cleanroom environments.
  • Reusable cleanroom glove
  • ANSI A7 cut resistance
  • Made in USA
Category III for particulates.
Certyfikaty: ANSI A7 level F cut resistance, ANSI Level 3 abrasion resistance.