Drukuj
Cleanroom gloves, polychloroprene, sterile, Biogel® Neotech
Dostawca: MOLNLYCKE HEALTH CARE
4450955 4450960 4450965 4450970 4450975 4450980 4450985 4450990
112-6329EA 2630 PLN
112-6329 112-6330 112-6331 112-6332 112-6333 112-6334 112-6335 112-6336
Cleanroom gloves, polychloroprene, sterile, Biogel® Neotech
Rękawice
These sterile cleanroom gloves with beaded cuffs and curved fingers are suitable for use in aseptic and Class 100 (ISO 5)/EU GMP Grade A environments. Recommended to be worn in combination with the Biogel® NeoTech Indicator® Underglove to form a Breach Indication System.
  • Can be worn together with Biogel® NeoTech Indicator® Underglove to form a Breach Indication System
  • Tested and cleared for use with chemotherapy agents
  • Synthetic polychloroprene
  • Biogel® hydrogel polymer coating for easy donning
  • Exceed industry standard for force at break, tensile strength, and elongation
  • Every glove (100%) is air-inflation tested for holes typically not detected in a visual inspection
Ideal for use in controlled environments, in medical and research laboratories, in the pharma industry, as well as in biotechnology.
Certyfikaty: EN 455, ASTM D3577, ISO 10282, EN 374, EN 16523-1, PPE Cat. III, AQL 0,65, Quality/Environment ISO 13485, ISO 14001, Sterilisation ISO 11137, Viral penetration ISO 16604, ASTM F1671, Allergenicity ISO 10993, Pyrogenicity ASTM D7102, Labelling EN 556-1, EN ISO 15223-1, EN ISO 21420.
Sposób pakowania: One pair per pack, in a high quality polyethylene inner wrap, packed into a film pack. 25 pairs per inner LDPE polybag f; 2 inner LDPE polybags are packed in an outer LDPE polybag. Four outer polybags per transit case, total of 200 pairs.