Drukuj
Cleanroom gloves, polyisoprene, sterile, Biogel® PI Tech
Dostawca: MOLNLYCKE HEALTH CARE
4440975 4440955 4440960 4440965 4440970 4440985 4440980 4440990
112-6358EA 3540 PLN
112-6358 112-6354 112-6355 112-6356 112-6357 112-6360 112-6359 112-6361
Cleanroom gloves, polyisoprene, sterile, Biogel® PI Tech
Rękawice
These sterile, cleanroom gloves with beaded anti-slip cuffs and curved fingers are suitable for use in aseptic and Class 100 (ISO 5)/EU GMP Grade A environments. Recommended to be worn in combination with the Biogel® PI Tech Indicator Underglove to form a Breach Indication System.
  • Can be worn together with Biogel® PI Tech Indicator® Underglove to form a Breach Indication System
  • Tested and cleared for use with chemotherapy agents
  • Synthetic polyisoprene
  • Biogel® hydrogel polymer coating for easy donning
  • Exceed industry standard for force at break, tensile strength, and elongation
  • Every glove (100%) is air-inflation tested for holes typically not detected in a visual inspection
Ideal for use in controlled environments, in medical and research laboratories, in the pharma industry, as well as in biotechnology.
Certyfikaty: EN 455, ASTM D3577, ISO 10282, EN 374, EN 16523-1, PPE Cat. III, AQL 0,65, Quality/Environment ISO 13485, ISO 14001, Sterilisation ISO 11137, Viral penetration ISO 16604, ASTM F1671, Allergenicity ISO 10993, Pyrogenicity ASTM D7102, Labelling EN 556-1, EN ISO 15223-1, EN ISO 21420.
Sposób pakowania: One pair per pack, in a high quality polyethylene inner wrap, packed into a film pack. 25 pairs per inner LDPE polybag f; 2 inner LDPE polybags are packed in an outer LDPE polybag. Four outer polybags per transit case, total of 200 pairs.