Drukuj
Klawiatura silikonowa dla sektora medycznego, GCQ CleanType® Easy Protect
Dostawca: GETT GERATETECHNIK
KG25240 KG25220 KG25219 KG25238 KG25242
GETTKG25240EA 700 PLN
GETTKG25240 GETTKG25220 GETTKG25219 GETTKG25238 GETTKG25242
Klawiatura silikonowa dla sektora medycznego, GCQ CleanType® Easy Protect
Klawiatury
Fully water-proof and dust-proof medical keyboard with classic layout.
  • The keyboard is IP 68 rated, which allows for fast, easy, and thorough cleaning
  • To avoid misusage during the cleaning process, keyboard buttons can be disabled by hitting the keylock button
  • Ergonomic design with a slightly reduced angle of 4° for comfortable operation
  • Shortened USB cable with protection cap
  • Two year warranty
  • Engineered in Germany
The CleanType® Easy Protect is a high-quality silicone keyboard that is easy to clean thanks to its water and dust resistance, making it ideal for use in hygiene-sensitive areas such as hospitals, clinics, medical practices, laboratories, or pharmaceutical companies.
Certyfikaty: CE, EMC, REACH, IP 68.
Informacje do zamówienia: Includes USB extension cable.
Sposób pakowania: The keyboards are packed in brown cardboard boxes.