Drukuj
VWR®, system poboru dla beczek ze stali nierdzewnej o pojemności 30 l, poziomy
Dostawca: VWR
VWR®
99901.030EA 2580 PLN
99901.030
VWR®, system poboru dla beczek ze stali nierdzewnej o pojemności 30 l, poziomy
Pompy Pompy do beczek
Specially developed solvent withdrawal system with manual pressure build-up for VWR® returnable 30 L stainless steel barrels.
  • Withdrawal system for 30 L barrels (stainless steel)
  • Perfluorelastomer o-rings
  • Includes 1 dip tube for 30 L barrels
This set contains a specially developed solvent withdrawal system for manual pressure build-up which can be used with VWR® 30 L returnable stainless steel barrels.