Drukuj
VWR®, system poboru dla beczek ze stali nierdzewnej o pojemności 10 l, poziomy
Dostawca: VWR
VWR®
99901.010EA 2540 PLN
99901.010
VWR®, system poboru dla beczek ze stali nierdzewnej o pojemności 10 l, poziomy
Pompy Pompy do beczek
Specially developed solvent withdrawal system with manual pressure build-up for VWR® returnable 10 L stainless steel barrels.
  • Withdrawal system for 10 L barrels (stainless steel)
  • Perfluorelastomer O-rings
  • Includes 1 dip tube for 10 L barrels
This set contains a specially developed solvent withdrawal system for manual pressure build-up which can be used with VWR® 10 L returnable stainless steel barrels.