Drukuj
Owijki do sterylizacji sekwencyjnej SMS, niebieskie, super wytrzymałe, 88 g/m²
Dostawca: KEYSTONE CLEANROOM PRODUCTS
KCRP-600-01.5 KCRP-600-0202 KCRP-600-0303 KCRP-600-0404 KCRP-600-0606 KCRP-600-0808 KCRP-600-0909 KCRP-600-0912 KCRP-600-1212 KCRP-600-1515 KCRP-600-1818 KCRP-600-1822 KCRP-600-2020 KCRP-600-2424 KCRP-600-3030 KCRP-600-3636 KCRP-600-4040 KCRP-600-4545 KCRP-600-4548 KCRP-600-4848 KCRP-600-5454 KCRP-600-5472 KCRP-600-6060 KCRP-600-6080
129-3775EA 3440 PLN
129-3775 129-3776 129-3777 129-3778 129-3779 129-3780 129-3781 129-3782 129-3783 129-3784 129-3785 129-3786 129-3787 129-3788 129-3789 129-3790 129-3791 129-3792 129-3793 129-3794 129-3795 129-3796 129-3797 129-3798
Owijki do sterylizacji sekwencyjnej SMS, niebieskie, super wytrzymałe, 88 g/m²
Folie do sterylizacji
Ideal for pharmaceutical and biotech production applications.
  • Manufactured and packaged in a certified ISO 8 cleanroom
  • Clean - no cellulose
  • Strong, rip and tear resistant 88 gsm SMS material
  • An extensive range of sizes available means less cutting
  • Hydrophobic barrier allows sterilisation gases, but prevents liquid and microbial penetration
  • Sub packed into sealed labelled poly bags with lot number and date of manufacture for complete traceability
For use in steam, ethylene oxide and hydrogen peroxide sterilisation processes.
Certyfikaty: Certificates of Conformance sent with shipments.
Sposób pakowania: The SMS wraps are packed in sealed, labelled poly bags with lot number and expiration date for traceability.