Drukuj
Wkładki do wiader z mopem, traktowane promieniami gamma, grubość 4 mm
Dostawca: KEYSTONE CLEANROOM PRODUCTS
S2828CP41 S2828CP41DP S2828CP42 S2828CP42DP S2828CP43 S2828CP43DP S3628CP41 S3628CP41DP S3628CP42 S3628CP42DP
129-2965EA 11140 PLN
129-2965 129-2966 129-2967 129-2968 129-2969 129-2970 129-2971 129-2972 129-2973 129-2974
Wkładki do wiader z mopem, traktowane promieniami gamma, grubość 4 mm
Worki Worki na odpady
Reduce the number of janitorial steam sterilisation loads by using gamma irradiated mop bucket liners. Instead of before every cleaning, the mop bucket system is typically autoclaved only once per week, but with each cleaning, a new sterile mop bucket liner pack is opened, the bucket is lined, and cleaning solution is added to it for cleaning.
  • Gamma irradiated, made from PE
  • Ready to use
  • Packaged in easy to open Tyvek peel pouches
  • Lowers labour costs
  • Reduces utility costs
  • Reduces washing/sanitisation/autoclaving of janitorial loads
  • Reduces exposure to chemicals and vapours
  • Ensures sterile receptacle and eliminates possibility of spreading mould spores from mop bucket
  • Simplifies disposal of cleaning solution
  • Ensures cleaning equipment is readily available
Gamma irradiated mop bucket liners instantly create a sterile receptacle for sterile cleaning solutions. Our rugged liners are supplied in 1, 2, 3 or 4 per pack, designed to accommodate the normal types of bucket systems in use in today's typical pharmaceutical production environments.
The use of a mop bucket liner eliminates the need to autoclave mop buckets after every use. After a cleaning process is complete, the liner and cleaning solution for is removed together for disposal. Since the inner surface of the bucket has not been in contact with the solution, it can remain in the clean storage area for the next use.
Certyfikaty: Certificates of irradiation and conformance with each shipment.