Drukuj
VWR®, butelki, HDPE
Dostawca: VWR Collection
215-4497EA 1250 PLN
215-4497 215-4498 215-4499 215-4504 215-4505 215-4506 215-4507 215-4508 215-4509 215-4510 215-4511 215-4512 215-4513 215-4514 215-4515 215-4516 215-4524 215-4525 215-4526 215-4527
VWR®, butelki, HDPE
Butelki i butle Butle plastikowe
Bottles are made with HDPE resin.
  • Sterile bottles feature an SAL of 10⁻³
  • Wide mouth design for easy filling
  • Available with multiple cap styles and pouring ring
  • Packed in smaller packs for better handling and to reduce contamination
Bottles feature graduation marks for reference during sample collection.
Certyfikaty: HDPE resin is in compliance with US FDA 21 CFR.
Sposób pakowania: Bottles are packed in shrink-wrapped modules.