Drukuj
Okulary ochronne, seria 4800
Dostawca: 3M
MMMABUD48AFEA 30.1 PLN
MMMABUD48AF
Okulary ochronne, seria 4800
Gogle
3M™ 4800 series safety goggles feature a clear, strong polycarbonate lens for impact resistance and indirect ventilation for added comfort.
  • Basic economical safety goggle with a flat lens design
  • Indirect ventilation to protect against liquid droplets
  • Polycarbonate lens for impact resistance
  • Not designed for wear over prescription glassess
  • Anti-fog coating to help reduce lens fogging
For all-round safety goggles which provide cooling comfort, the 4800 Series features an indirect ventilation system to prevent misting and ensures comfort while working over long periods. They also have a clear, strong polycarbonate lens to protect against impacts and flying objects. They are available with an anti-fog lens coating.