Drukuj
Cylindryczne złącza proste z zadziorami, PVDF
Dostawca: Avantor Fluid Handling
06044-01 06044-04
MFLX06044-01EA 52.8 PLN
MFLX06044-01 MFLX06044-04
Cylindryczne złącza proste z zadziorami, PVDF
Okucia i złącza
Safe and flexible connection for your most demanding applications.
  • Withstands temperatures up to 320 °F (160 °C)
  • Highly resistant to chemicals and UV
  • Autoclavable
The cylindrical barbed straight connectors are available in a variety of sizes to extend, connect, branch, join, and repair most plastic tubings. The polyvinylidene fluoride (PVDF) connectors are designed for quick and easy assembly without any tools.