Drukuj
Buty ochronne ESD, sznurowane, MADDOX Air Mesh Low, O1
Dostawca: ELTEN
972560/36 972560/37 972560/38 972560/39 972560/40 972560/41 972560/42 972560/43 972560/44 972560/45 972560/46 972560/47 972560/48 972560/49
ELTN972560/36EA 501 PLN
ELTN972560/36 ELTN972560/37 ELTN972560/38 ELTN972560/39 ELTN972560/40 ELTN972560/41 ELTN972560/42 ELTN972560/43 ELTN972560/44 ELTN972560/45 ELTN972560/46 ELTN972560/47 ELTN972560/48 ELTN972560/49
Buty ochronne ESD, sznurowane, MADDOX Air Mesh Low, O1
Obuwie i buty
Excellent wearing comfort, with a padded upper edge to protect the Achilles tendon.
  • Microfibre/Mesh material
  • Fabric lining for natural ventilation
  • Padded tongue
  • Full-length inlay sole SPORTIVE ESD grey
  • Soft fleece ESD insole
  • TPU/PU sole MAXXIMO
  • Without protective toe cap
Thanks to its excellent discharge capability, the shoe is suitable for work in ESD sensitive or electrostatically protected areas (EPA). Sole core made of Infinergy® by BASF. Metal- and leather-free.
Certyfikaty: EN ISO 20347 O1 SRC, form A