Drukuj
Tagi
Dostawca: GA
CID-02WH CID-02YE
GAINCID-02WHEA 203 PLN
GAINCID-02WH GAINCID-02YE
Tagi
Etykiety
These tags are used for identification of metal racks and other containers for long-term storage in liquid nitrogen and ultra-low temperature freezers.
  • Suitable temperature ranges from –196 to +80 °C
Labels can be printed with all major brands of thermal transfer barcode printers (SATO, Zebra, Intermec, Printronix, Datamax-O'Neil, CAB, and more) or inscribed with permanent markers such as our cryo-markers.
Dodatkowe informacje: Includes 12 plastic Cryo-Cards™ with connection hooks and thermal transfer cryo labels.