Drukuj
Etykiety do głębokiego mrożenia dla drukarek laserowych
Dostawca: GA International
GAINHCL-10T1-REEA 334 PLN
GAINHCL-10T1-RE GAINHCL-10T1-LA GAINHCL-10T1-SI GAINHCL-10T1-PI GAINHCL-10T1-LE GAINHCL-10T1-BL GAINHCL-10T1-YE GAINHCL-10T1-WH GAINHCL-10T1-A1 GAINHCL-10T1-BR GAINHCL-10T1-OR GAINHCL-10T1-MT GAINHCL-10T1-GA
Etykiety do głębokiego mrożenia dla drukarek laserowych
Etykiety
Deep-freeze thermoplastic laser labels for long-term storage in –80 °C freezers.
  • Available in a wide range of colours; perfect for colour-coding
  • Labels withstand sterilization by steam, dry heat, and gamma irradiation
  • Strong permanent adhesive ensures labels won't wrinkle or peel
  • Printable by all major desktop laser printers
  • Resistant to gamma irradiation (tested to up to 50 kGy)
These circular labels are provided in a range of colours and accept laser printing and writing with permanent cryo markers. Designed for the identification of 0,5, 1,5, and 2 ml microtube and vial caps. Water-resistant, wear and tear proof, these labels will not jam printers. They are resistant to autoclaving, UV, and Gamma irradiation.