Drukuj
Buty ochronne ESD, sznurowane, Marten XXSports Low
Dostawca: ELTEN
728160/35 728160/36 728160/37 728160/38 728160/39 728160/40 728160/41 728160/42 728160/43 728160/44 728160/45 728160/46 728160/47 728160/48
ELTN728160/35EA 496 PLN
ELTN728160/35 ELTN728160/36 ELTN728160/37 ELTN728160/38 ELTN728160/39 ELTN728160/40 ELTN728160/41 ELTN728160/42 ELTN728160/43 ELTN728160/44 ELTN728160/45 ELTN728160/46 ELTN728160/47 ELTN728160/48
Buty ochronne ESD, sznurowane, Marten XXSports Low
Obuwie i buty
Thanks to its excellent discharge capability, the shoe is suitable for work in ESD sensitive or electrostatically protected areas (EPA).
  • Full, padded bellows tongue
  • TPU/PU sole, WELLMAXX SPORTS
  • Steel toe cap
  • Leather-free
Mesh material/microfibre construction with fabric lining for natural ventilation. Full-length inlay sole, SPORTIVE ESD. Anti-static soft fleece insole. Lacing system with individual quick locking system. Sole core made of Infinergy® by BASF.
Colour: Grey/red
Certyfikaty: EN ISO 20345 S1 SRA, Form A.