Drukuj
Single element standard for AA and MP-AES, Thallium
Dostawca: Agilent
5190-8316 5190-8317
HEWL5190-8316EA 222 PLN
HEWL5190-8316 HEWL5190-8317
Single element standard for AA and MP-AES, Thallium
Standardy Wzorce kationowe Thallium Standards
Agilent offers a comprehensive range of single and multi-element standards, ionization buffers and environmental standards for Atomic Spectroscopy.
  • Gives you confidence in your AAS, ICP-OES or ICP-MS and MP-AES results
  • Accurate, reliable calibrations
All of Agilent's standards are prepared to exacting specifications that ensure consistency. Every solution is manufactured using high purity starting materials, which are regularly tested in accordance with NIST guidelines to provide traceability.
Dodatkowe informacje: Supplied with a Material Safety Data Sheet (MSDS) and Certificate of Analysis for complete assurance.
Sposób pakowania: Packaged in a pre-leached HDPE bottle, secured with a tamper-evident seal.