Drukuj
Single element standards for ICP-OES, thulium
Dostawca: Agilent
5190-8541 5190-8542 5190-8219 5190-8220
HEWL5190-8541EA 717 PLN
HEWL5190-8541 HEWL5190-8542 HEWL5190-8219 HEWL5190-8220
Single element standards for ICP-OES, thulium
Standardy Wzorce kationowe Thulium Standards
They offer a comprehensive range of single and multi-element standards, tuning solutions, ionisation buffers and environmental standards for Atomic Spectroscopy.
  • Gives you confidence in our AAS, ICP-OES or ICP-MS and MP-AES results
  • Accurate, reliable calibrations
All of standards are prepared to exacting specifications that ensure consistency. Every solution is manufactured using high purity starting materials, which are regularly tested in accordance with NIST guidelines to provide traceability.
Dodatkowe informacje: Supplied with a Material Safety Data Sheet (MSDS) and Certificate of Analysis for complete assurance.
Sposób pakowania: Packaged in a pre-leached HDPE bottle, secured with a tamper-evident seal.