Drukuj
Single element standard for AA and MP-AES, Nickel
Dostawca: Agilent
5190-8298 5190-8299
HEWL5190-8298EA 181 PLN
HEWL5190-8298 HEWL5190-8299
Single element standard for AA and MP-AES, Nickel
Standardy Wzorce kationowe Nickel Standards
They offer a comprehensive range of single and multi-element standards, ionisation buffers and environmental standards for Atomic Spectroscopy.
  • Gives you confidence in your AAS, ICP-OES or ICP-MS and MP-AES results
  • Accurate, reliable calibrations
All of standards are prepared to exacting specifications that ensure consistency. Every solution is manufactured using high purity starting materials, which are regularly tested in accordance with NIST guidelines to provide traceability.
Dodatkowe informacje: Supplied with a Material Safety Data Sheet (MSDS) and Certificate of Analysis for complete assurance.
Sposób pakowania: Packaged in a pre-leached HDPE bottle, secured with a tamper-evident seal.