Drukuj
Materiały eksploatacyjne do próbników headspace
Dostawca: Agilent
RDT-1020 G3520-20210 0100-2595 G4556-60019 G4556-67010 G4556-80101 G4556-80102 G4556-80103 G4556-80105 G4556-80106 G4556-80108 G4556-80109 G4556-80126 G4556-80128 G4556-90500 4177-0607 G3452-60835 G4556-80107
HEWLRDT-1020EA 1380 PLN
HEWLRDT-1020 HEWLG3520-20210 HEWL0100-2595 HEWLG4556-60019 HEWLG4556-67010 HEWLG4556-80101 HEWLG4556-80102 HEWLG4556-80103 HEWLG4556-80105 HEWLG4556-80106 HEWLG4556-80108 HEWLG4556-80109 HEWLG4556-80126 HEWLG4556-80128 HEWLG4556-90500 HEWL4177-0607 HEWLG3452-60835 HEWLG4556-80107
Materiały eksploatacyjne do próbników headspace
Materiały zużywalne do chromatografii Części zamienne do chromatografów
Find the headspace supplies you need for your Agilent 7697 or G1888 headspace sampler.
  • Built-in pneumatics for superior control and easier setup
  • Proven valve and loop sampling technology
  • Fully-automatic sample vial leak checking and available bar code reader help ensure greater confidence in results method compatibility
  • Instrument control software that is fully integrated in data systems
  • Resource conserving programmable instrument scheduler
Headspace sampling allows introduction of volatile compounds to the GC or GC/MS from virtually any sample matrix while leaving unwanted components in a disposable sample vial. This results in less maintenance and longer lifetimes for the GC consumables in your lab. With up to 111 sample vial positions and removable vial racks, the 7697A's inert sample pathway supports nearly continuous operation to satisfy even the busiest laboratory. Alternatively, it's legacy G1888 headspace sampler also features an inert sample pathway for superior chemical performance without analyte degradation or loss.