Drukuj
Płyty filtracyjne Captiva
Dostawca: Agilent
A5962012 A5967045
HEWLA5962012EA 3490 PLN
HEWLA5962012 HEWLA5967045
Płyty filtracyjne Captiva
Mikropłytki Płytki filtracyjne
Filtration is simple, versatile, and necessary to prevent clogging of HPLC columns. Captiva's unique dual depth filtration media provides complete removal of precipitated proteins and outstanding resistance to sample clogging.
  • Standard SPE format
  • Ideal for LC/MS samples
  • Avoid sample transfer problems
  • Nondrip 3 ml cartridges resist flow until vacuum is applied
  • Captiva ND Lipids effectively remove ion suppressing phospholipids from biological samples
  • Protect HPLC columns from clogging to reduce instrument downtime
  • Clean and clear filtrates offer improved sensitivity
Filtration is simple, versatile, and necessary to prevent clogging of HPLC columns. Captiva's unique dual depth filtration media provides complete removal of precipitated proteins and outstanding resistance to sample clogging. Captiva 96-well filter plates are available in 0,2 and 0,45 µm depths, and are ideal for preparing precipitated protein samples for LC/MS.
Captiva 10 and 20 µm glass fiber filter plates are designed for clarifying highly particle-laden samples. These plates are perfect for automated systems and for use with Duo Seal 96-well seals. All Captiva components are ultra clean, and rigorously tested to ensure against nonspecific binding.
Dodatkowe informacje: Starter kits contain everything you need to get up and running with minimum fuss. Replacement kits include everything you need to replenish your captiva system.
Uwaga: For research use only. Not for use in diagnostic procedures.