Drukuj
Multiple distributor caps for drums with GL thread connections, BOLA
Dostawca: Bohlender
D698-14 D696-08 D696-14 D698-08 D697-14 D697-08
216-0570EA 1870 PLN
216-0570 216-0567 216-0568 216-0569 216-1741 216-1740
Multiple distributor caps for drums with GL thread connections, BOLA
Nakrętki Nakrętki
PTFE.
  • Very good chemical resistance, the fluid medium only comes into contact with PTFE
  • Temperature resistant from −200 to +250 °C
  • Pressure resistant up to a maximum of 1 bar, vacuum suitable
Screw cap made from glass fibre reinforced PTFE, suitable for drums/barrels with internal thread; movable distributor body with GL connector necks made from PTFE. For the removal and filling of aggressive or pure liquids and insertion of hoses, tubes and probes into drums.