Drukuj
Zestaw do pisania kriogenicznego, Nunc™
Dostawca: Bohlender
A117-04 A117-08
201-0014EA 2670 PLN
201-0014 201-0009
Zestaw do pisania kriogenicznego, Nunc™
Butelki i butle Płuczki gazowe
Base, upper head connections and frit made from PTFE, column body, riser, inlet and outlet tubes made from FEP.
  • Very good chemical resistance
  • Temperature resistant from −200 to +205 °C
  • Transparent, unbreakable
Intensive gas exchange due to large riser height. The standard frit made from porous PTFE (approximately 3 µm pore size) is screwed to the riser (thread M8×1) and can therefore be easily exchanged.