Drukuj
Single element standards for ICP-OES, niobium
Dostawca: Agilent
5190-8424 5190-8425 5190-8493 5190-8494
HEWL5190-8424EA 1010 PLN
HEWL5190-8424 HEWL5190-8425 HEWL5190-8493 HEWL5190-8494
Single element standards for ICP-OES, niobium
Standardy Wzorce kationowe Niobium Standards
Agilent offers a comprehensive range of single and multi-element standards, tuning solutions, ionisation buffers and environmental standards for Atomic Spectroscopy.
  • Gives you confidence in our AAS, ICP-OES or ICP-MS and MP-AES results
  • Accurate, reliable calibrations
All of Agilent's standards are prepared to exacting specifications that ensure consistency. Every solution is manufactured using high purity starting materials, which are regularly tested in accordance with NIST guidelines to provide traceability.
Dodatkowe informacje: Supplied with a Material Safety Data Sheet (MSDS) and Certificate of Analysis for complete assurance.
Sposób pakowania: Packaged in a pre-leached HDPE bottle, secured with a tamper-evident seal.