Drukuj
Single element standard for AA and MP-AES, Aluminium
Dostawca: Agilent
5190-8256 5190-8257
HEWL5190-8256EA 208 PLN
HEWL5190-8256 HEWL5190-8257
Single element standard for AA and MP-AES, Aluminium
Standardy Wzorce kationowe Wzorce glinu
Agilent offers a comprehensive range of single and multi-element standards, ionisation buffers and environmental standards for Atomic Spectroscopy.
  • Gives you confidence in your AAS, ICP-OES or ICP-MS and MP-AES results
  • Accurate, reliable calibrations
All of Agilent's standards are prepared to exacting specifications that ensure consistency. Every solution is manufactured using high purity starting materials, which are regularly tested in accordance with NIST guidelines to provide traceability.
Dodatkowe informacje: Supplied with a Material Safety Data Sheet (MSDS) and Certificate of Analysis for complete assurance.
Sposób pakowania: Packaged in a pre-leached HDPE bottle, secured with a tamper-evident seal.