Drukuj
Matrix modifiers for graphite furnace AA
Dostawca: Agilent
5190-8335 5190-8336 5190-8337 5190-8338 5190-8339 5190-8340 5190-8341 5190-8342
HEWL5190-8335EA 905 PLN
HEWL5190-8335 HEWL5190-8336 HEWL5190-8337 HEWL5190-8338 HEWL5190-8339 HEWL5190-8340 HEWL5190-8341 HEWL5190-8342
Matrix modifiers for graphite furnace AA
Standardy Wzorce kationowe Multi-Element Standards
Agilent offers a comprehensive range of single and multi-element standards, ionisation buffers and environmental standards for Atomic Spectroscopy.
  • Gives you confidence in your AAS, ICP-OES or ICP-MS and MP-AES results
  • Accurate, reliable calibrations
All of Agilent's standards are prepared to exacting specifications that ensure consistency. Every solution is manufactured using high purity starting materials, which are regularly tested in accordance with NIST guidelines to provide traceability.
Dodatkowe informacje: Supplied with a Material Safety Data Sheet (MSDS) and Certificate of Analysis for complete assurance.
Sposób pakowania: Packaged in a pre-leached HDPE bottle, and secured with a tamper-evident seal.