Drukuj
Kolumienki GC, J&W DB-1ht
Dostawca: Agilent
122-1111 122-1131 123-1111 123-1131
HEWL122-1111EA 3640 PLN
HEWL122-1111 HEWL122-1131 HEWL123-1111 HEWL123-1131
Kolumienki GC, J&W DB-1ht
Kolumny chromatograficzne
DB-1ht are specially processed for extended temperature limits of 400 °C with excellent peak shape and faster elution times for high boilers.
  • 100% Dimethylpolysiloxane
  • Non-polar
  • High temperature, polyimide-coated, fused silica tubing
  • Excellent peak shape and faster elution times for high boilers
  • Faster elution times for high boiling compounds and are solvent rinsable
Agilent JW DB-1ht delivers excellent performance in a range of GC/MS applications and are manufactured to the highest levels of quality and stability. Nonpolar DB-1ht columns are specially processed for operation up to 400 °C, with optimised, polyimide-coated fused silica tubing.