Drukuj
Szybkozłączne moduły palnika dla PerkinElmer
Dostawca: Agilent
8003-0336 8003-0337 8003-0338 8003-0339 8003-0340 8003-0342 8003-0343
HEWL8003-0336EA 31920 PLN
HEWL8003-0336 HEWL8003-0337 HEWL8003-0338 HEWL8003-0339 HEWL8003-0340 HEWL8003-0342 HEWL8003-0343
Szybkozłączne moduły palnika dla PerkinElmer
Materiały zużywalne do chromatografii Części zamienne do chromatografów
The Quick-Connect torch module for PerkinElmer ICP-OES includes the torch, mounting block, 2 mm alumina injector, Scott spray chamber, and Cross-Flow nebuliser.
  • Guaranteed to be compatible with PerkinElmer instruments
  • Backed by the stand behind warranty that guarantees the parts will not cause instrument failure or downtime
  • Parts qualified on a genuine PerkinElmer system
  • Many parts in packaging designed to be more durable, sturdy and provide the best protection during shipping to ensure the part arrives with no damage
The Quick-Change module is useful when you are running different sample types and need separate sample introduction for each.