Drukuj
Single element standards for ICP-OES, molybdenum
Dostawca: Agilent
5190-8418 5190-8419 5190-8487 5190-8488
HEWL5190-8418EA 771 PLN
HEWL5190-8418 HEWL5190-8419 HEWL5190-8487 HEWL5190-8488
Single element standards for ICP-OES, molybdenum
Standardy Wzorce kationowe Molybdenum Standards
Agilent offers a comprehensive range of single and multi-element standards, tuning solutions, ionisation buffers and environmental standards for Atomic Spectroscopy.
  • Gives you confidence in our AAS, ICP-OES or ICP-MS and MP-AES results
  • Accurate, reliable calibrations
All of Agilent's standards are prepared to exacting specifications that ensure consistency. Every solution is manufactured using high purity starting materials, which are regularly tested in accordance with NIST guidelines to provide traceability.
Dodatkowe informacje: Supplied with a Material Safety Data Sheet (MSDS) and Certificate of Analysis for complete assurance.
Sposób pakowania: Packaged in a pre-leached HDPE bottle, secured with a tamper-evident seal.