Drukuj
Single element standards for ICP-OES, rhodium
Dostawca: Agilent
5190-8509 5190-8510
HEWL5190-8509EA 2320 PLN
HEWL5190-8509 HEWL5190-8510
Single element standards for ICP-OES, rhodium
Standardy Wzorce kationowe Rhodium Standards
Agilent offers a comprehensive range of single and multi-element standards, tuning solutions, ionisation buffers and environmental standards for Atomic Spectroscopy.
  • Gives you confidence in our AAS, ICP-OES or ICP-MS and MP-AES results
  • Accurate, reliable calibrations
All of Agilent's standards are prepared to exacting specifications that ensure consistency. Every solution is manufactured using high purity starting materials, which are regularly tested in accordance with NIST guidelines to provide traceability.
Dodatkowe informacje: Supplied with a Material Safety Data Sheet (MSDS) and Certificate of Analysis for complete assurance.
Sposób pakowania: Packaged in a pre-leached HDPE bottle, secured with a tamper-evident seal.