Drukuj
Lipaza Rhizopus oryzae
Dostawca: Apollo Scientific
BIIM1003-10G BIIM1003-25G BIIM1003-50G
APOSBIIM1003-10GEA 1420 PLN
APOSBIIM1003-10G APOSBIIM1003-25G APOSBIIM1003-50G
Lipaza Rhizopus oryzae
Enzymy
Covalent on IB-150A. Activity: 500 TBU/g
  • immobilized on Immobead 150