Drukuj
Aprotinin
Dostawca: Apollo Scientific
BIMI2132-100MG BIMI2132-250MG BIMI2132-25MG
APOSBIMI2132-100MGEA 1210 PLN
APOSBIMI2132-100MG APOSBIMI2132-250MG APOSBIMI2132-25MG
Aprotinin
Białka
Aprotinin inhibits trypsin, chymotrypsin, kallikrein from different sources and plasmin.