Drukuj
Autoklawowalne rurki do ciągłego formowania, Tyvek® 1422A
Dostawca: KEYSTONE CLEANROOM PRODUCTS
TUBT2A02 TUBT2A03 TUBT2A04 TUBT2A05 TUBT2A06 TUBT2A08 TUBT2A10 TUBT2A12 TUBT2A15 TUBT2A16 TUBT2A24 TUBT2A28 TUBT2A30 TUBT2A40
129-2996EA 11280 PLN
129-2996 129-2997 129-2998 129-2999 129-3001 129-3002 129-3003 129-3004 129-3005 129-3006 129-3007 129-3008 129-3009 129-3012
Autoklawowalne rurki do ciągłego formowania, Tyvek® 1422A
Worki Worki do sterylizacji
Continuous form tubing made from 1422A Tyvek® provides the user with tremendous sizing flexibility when wrapping pharmaceutical equipment for steam sterilisation.
  • Manufactured and packaged in a controlled cleanroom environment
  • On a plastic core (no corrugated material)
  • Tear and puncture resistant
  • Hydrophobic
  • Lint-free
  • Consistently strong factory seals
  • Exceptional microbial barrier
1422A Tyvek® steam sterilisation tubing is a great choice when there is a wide range of equipment parts that need to be wrapped for sterilization.
Simply unroll the tubing to the desired length, slide the equipment into the tube and then heat seal each end closed.
The drapeable nature of the 1422A Tyvek® material is ideal for wrapping equipment parts where one may need to 'feel through' the bag; especially when lifting, or handling the wrapped part.
Since 100% of the surface area allows for steam penetration and drying, each load can be configured for maximum efficiency and throughput.
Dodatkowe informacje: Certificates of conformance sent with each shipment.