Drukuj
Złącza rurek Y, b.safe
Dostawca: Bohlender
M580-02 M580-04 M580-06 M580-08 M580-10 M580-12
590-0222EA 45 PLN
590-0222 590-0223 590-0224 590-0225 590-0226 590-0227
Złącza rurek Y, b.safe
Okucia i złącza
Y-shaped fitting made of PP, with three hose connectors for elastic tubing.
  • Excellent chemical resistance
  • Available for a wide range of hose diameters
  • The conical shape gets the connection tight and leak-free
b.safe hose connectors are the flexible choice to customise your b.safe waste cap. Made of PP or PTFE and available in numerous diameters you will find the exact one to fit your waste hose.