Drukuj
Adaptery do podłączenia filtra wydechowego, b.safe
Dostawca: Bohlender
M516-01 M516-05
590-0202EA 138 PLN
590-0202 590-0203
Adaptery do podłączenia filtra wydechowego, b.safe
Okucia i złącza
Adaptor for installation of capillaries with b.safe fittings at the exhaust filter connection of the b.safe waste caps.
  • Excellent chemical resistance
  • Converts the exhaust filter port into a capillary port
  • Suitable for different capillary diameters
Suitable b.safe fittings for different capillary diameters are separately available.