Drukuj
Adaptery do filtrów wylotowych, b.safe
Dostawca: Bohlender
M502-01 M502-02 M502-03 M502-04 M615-20
590-0158EA 155 PLN
590-0158 590-0159 590-0160 590-0161 590-0247
Adaptery do filtrów wylotowych, b.safe
Okucia i złącza
Adaptor for space saving installation of the b.safe exhaust filter on b.safe waste caps.
  • Enlarge the space on b.safe waste caps
  • Reduce the installation height
  • space saving
Enlarge the space on b.safe waste caps with b.safe adaptors for exhaust filters and facilitate the installation of capillaries and tubing. With an adapter 1×90°, cat. no. VWRI590-0160, you reduce the installation height of the waste cap with mounted exhaust filter on your canister.