Drukuj
Vialki z szyjką zaciskową, La-Pha-Pack™, ND8
Dostawca: LA PHA PACK
08090405 08090406
548-3036EA 275 PLN
548-3036 548-3037
Vialki z szyjką zaciskową, La-Pha-Pack™, ND8
Vialki
These vials are for use on instruments from the following manufacturers: Agilent, Beckman, Carlo Erba, CTC, Fisons, PerkinElmer, Shimadzu, Thermo Scientific, VWR®/Hitachi, etc.
  • Flat, round or conical bottom
  • In clear or amber glass
  • For almost all autosamplers
Broad selection of crimp neck vials ND8 available: Crimp neck vials and micro-vials ND8 can be closed with 8 mm aluminium caps, 9 mm PE caps or with 8 mm push-on caps. However, micro-vials often need an adapter to run in the autosampler. They often have a conical bottom shape, so need an adapter to stand.