Drukuj
Rękawice chemoodporne, Sandy Grip Lite, AP60
Dostawca: Portwest
PRTWAP60B8RXXLEA 847 PLN
PRTWAP60B8RXXL PRTWAP60B8RM PRTWAP60B8RL PRTWAP60B8RXL
Rękawice chemoodporne, Sandy Grip Lite, AP60
Rękawice
Nitrile gauntlet with sandy palm dip for a secure grip and abrasion resistance. This glove is designed for use in wet and oil environments. Using a lightweight soft comfortable liner, it increases the wearability for prolonged periods of time.
  • Nitrile sandy coating
  • Lightweight and comfortable
  • 13 gauge liner for a perfect fit
  • 62 mil / 1,6 mm enhanced thickness on the palm for added durability
Oil and water resistant. Flexible sandy nitrile coating offers great grip in wet and dry conditions. Designed with a comfort fit.
Materials: Nylon, nitrile, nitrile Sandy
Certyfikaty: AS/NZS 2161 .2, AS/NZS 2161 .3, EN ISO 374-5 Micro Organisms Pass, EN 420, EN388:2016 +A1:2018 - 4.1.2.1.X., EN ISO 374-1:2016 Type B A.J.K.L.