Drukuj
Buty robocze, sznurowane, FW19
Dostawca: Portwest
FW19BKR36 FW19BKR37 FW19BKR38 FW19BKR39 FW19BKR40 FW19BKR41 FW19BKR42 FW19BKR43 FW19BKR44 FW19BKR45 FW19BKR46 FW19BKR47 FW19BKR48
PRTWFW19BKR36EA 192 PLN
PRTWFW19BKR36 PRTWFW19BKR37 PRTWFW19BKR38 PRTWFW19BKR39 PRTWFW19BKR40 PRTWFW19BKR41 PRTWFW19BKR42 PRTWFW19BKR43 PRTWFW19BKR44 PRTWFW19BKR45 PRTWFW19BKR46 PRTWFW19BKR47 PRTWFW19BKR48
Buty robocze, sznurowane, FW19
Obuwie i buty
Lightweight and flexible work shoe with excellent durability. Dual density, oil resistant and energy absorbing seat region for lasting comfort.
  • Anti-static footwear
  • Slip resistant outsole (SRA)
  • Heat resistant outsole - 200
  • Oil resistant outsole
Material upper: split leather
Material outsole: PU
Certyfikaty: EN ISO 20347:2012 O1