Drukuj
Rękawice odporne na przecięcia, eco-cut, A635
Dostawca: Portwest
A635K8RM A635K8RL A635K8RS A635K8RXL
PRTWA635K8RMEA 295 PLN
PRTWA635K8RM PRTWA635K8RL PRTWA635K8RS PRTWA635K8RXL
Rękawice odporne na przecięcia, eco-cut, A635
Rękawice
These Eco-Cut gloves are conforming to the level B cut resistance and are ideal for tasks where the risk of injury is low. Features all the staple benefits of comfort, dexterity and grip.
  • 13-gauge liner for a perfect fit
  • Retail tag which aids presentation for retail sales
  • Low linting construction for minimal contamination
Materials: polyester, glass fibre, elastane, PU.
Certyfikaty: AS/NZS 2161 .2; AS/NZS 2161 .3; EN388:2016 +A1:2018 - 4.X.4.2.B; EN 420.