Drukuj
Base pro antibacterial leggings, B151
Dostawca: Portwest
B151BKRS B151BKRL B151BKRXL B151BKRXXL B151BKRM
PRTWB151BKRSEA 139 PLN
PRTWB151BKRS PRTWB151BKRL PRTWB151BKRXL PRTWB151BKRXXL PRTWB151BKRM
Base pro antibacterial leggings, B151
Odzież robocza
These leggings offers enhanced thermal protection, wicks moisture away from the body and combats odour, keeping the wearer cool, dry and comfortable under exerting conditions.
  • Designed with a comfort fit
  • Drying & wicking
  • Contrast colour stitching for added style
Material used is silver ion 165 g.