Drukuj
Rękawice spawalnicze TIG, A520
Dostawca: Portwest
A520GRRL A520GRRXXL A520GRRXL
PRTWA520GRRLEA 70.2 PLN
PRTWA520GRRL PRTWA520GRRXXL PRTWA520GRRXL
Rękawice spawalnicze TIG, A520
Rękawice
Cow split leather gloves with flexible goatskin palm and heavy duty split leather cuff. For use in type B (Tig Welding) applications.
  • Stitching made from para-aramid throughout
  • Ultimate dexterity for welders
Ideal for welding and metal handling.
Certyfikaty: EN 388:2016 (2122X), EN 407 (413X3X), EN 12477 Type B