Drukuj
Flame-retardant high visibility jacket, Bizflame™ Rain, S778
Dostawca: Portwest
S778ORR4XL S778ORRL S778ORRM S778ORRS S778ORRXL S778ORRXS S778ORRXXL S778ORRXXXL S778YERXXL S778YERXXXL S778YER4XL S778YER5XL S778YER6XL S778YERM S778YERS S778YERXS S778YERL S778YERXL
PRTWS778ORR4XLEA 1010 PLN
PRTWS778ORR4XL PRTWS778ORRL PRTWS778ORRM PRTWS778ORRS PRTWS778ORRXL PRTWS778ORRXS PRTWS778ORRXXL PRTWS778ORRXXXL PRTWS778YERXXL PRTWS778YERXXXL PRTWS778YER4XL PRTWS778YER5XL PRTWS778YER6XL PRTWS778YERM PRTWS778YERS PRTWS778YERXS PRTWS778YERL PRTWS778YERXL
Flame-retardant high visibility jacket, Bizflame™ Rain, S778
Odzież robocza
Multi-norm jacket made of 100% polyester flame with resistant filling. Flame resistance coupled with superb breathable and water resistant properties
  • Anti-static
  • Breathable, windproof and water resistant
  • Pack away hood
  • Two-way zipper
  • Triple stitched throughout for strength
Weight (outer fabric): 250 g/m²
Weight: (lining): 170 g/m²
Weight (fleece gilet): 350 g/m²
Industrial wash flame resistant tape provides enhanced visibility. Prevents build up of wearer static energy.
Certyfikaty: EN 1149 -5, EN ISO 14116 Index 3 (/12H/40), EN 13034 type 6, EN ISO 20471 Class 3 GO/RT 3279 Issue 8 (orange only), EN 343 Class 3:1
Uwaga: Use non-biological powder. Do not use softener.